EMAIL THIS PAGE TO A FRIEND
298638 Sigma-Aldrich

Hexadecanophenone

95%

Purchase

Properties

assay   95%
bp   248 °C/12 mmHg (lit.)
mp   61-65 °C (lit.)
SMILES string   CCCCCCCCCCCCCCCC(=O)c1ccccc1
InChI   1S/C22H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-17-20-22(23)21-18-15-14-16-19-21/h14-16,18-19H,2-13,17,20H2,1H3
InChI key   IIOLAWJMOGLOIB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?