• USA Home
  • 298999 - 1,1,1-Trichloroethane

EMAIL THIS PAGE TO A FRIEND
298999 Sigma-Aldrich

1,1,1-Trichloroethane

anhydrous, ≥99.5%, contains 0.05% low alkyl epoxide as stabilizer

Synonym: ‘Chlorothene’, Methylchloroform

  • CAS Number 71-55-6

  • Linear Formula Cl3CCH3

  • Molecular Weight 133.40

  •  Beilstein/REAXYS Number 1731614

  •  EC Number 200-756-3

  •  MDL number MFCD00000806

  •  PubChem Substance ID 24858006

Purchase

Properties

grade   anhydrous
vapor density   4.6 (vs air)
vapor pressure   100 mmHg ( 20 °C)
InChI Key   UOCLXMDMGBRAIB-UHFFFAOYSA-N
assay   ≥99.5%
autoignition temp.   998 °F
contains   0.05% low alkyl epoxide as stabilizer
expl. lim.   7.5-15 %, 25 °F
impurities   <0.005% water
evapn. residue   <0.0003%
refractive index   n20/D 1.4366 (lit.)
bp   74-76 °C(lit.)
mp   −35 °C (lit.)
density   1.336 g/mL at 20 °C (lit.)

Description

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2831 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

Use the interactive controls above to simplify calculations and improve the efficiency of your distillation or evaporation requirements.
Keywords: Evaporation, Vaporization

Properties of Solvents Table

The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?