• USA Home
  • 299731 - 2′-Fluoroacetanilide

EMAIL THIS PAGE TO A FRIEND
299731 Sigma-Aldrich

2′-Fluoroacetanilide

98%

Purchase

Properties

assay   98%
bp   140-142 °C/14 mmHg (lit.)
mp   77-79 °C (lit.)
SMILES string   CC(=O)Nc1ccccc1F
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?