• USA Home
  • 299871 - 2-Acetamido-7-fluorofluorene

EMAIL THIS PAGE TO A FRIEND
299871

2-Acetamido-7-fluorofluorene

98%

  • CAS Number 343-89-5

  • Empirical Formula (Hill Notation) C15H12FNO

  • Molecular Weight 241.26

  •  MDL number MFCD00010773

Purchase

Properties

assay   98%
SMILES string   CC(=O)Nc1ccc-2c(Cc3cc(F)ccc-23)c1
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?