• USA Home
  • 301205 - 1,2,3,4-Tetrachlorobenzene

EMAIL THIS PAGE TO A FRIEND
301205 Aldrich

1,2,3,4-Tetrachlorobenzene

technical grade, 90%

  • CAS Number 634-66-2

  • Empirical Formula (Hill Notation) C6H2Cl4

  • Molecular Weight 215.89

  •  Beilstein Registry Number 1910025

  •  EC Number 211-214-0

  •  MDL number MFCD00000538

  •  PubChem Substance ID 24858172

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
grade   technical grade
InChI Key   GBDZXPJXOMHESU-UHFFFAOYSA-N
assay   90%
bp   254 °C/761 mmHg(lit.)
mp   42-47 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?