• USA Home
  • 301442 - Hexyltriphenylphosphonium bromide

EMAIL THIS PAGE TO A FRIEND
301442 Sigma-Aldrich

Hexyltriphenylphosphonium bromide

95%

Purchase

Properties

assay   95%
mp   201-204 °C (lit.)
SMILES string   [Br-].CCCCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?