• USA Home
  • 302236 - 1,2-Dibromo-3,5-difluorobenzene

EMAIL THIS PAGE TO A FRIEND
302236

1,2-Dibromo-3,5-difluorobenzene

99%

  • CAS Number 139215-43-3

  • Empirical Formula (Hill Notation) C6H2Br2F2

  • Molecular Weight 271.88

  •  MDL number MFCD00042559

Purchase

Properties

assay   99%
SMILES string   Fc1cc(F)c(Br)c(Br)c1
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?