• USA Home
  • 302287 - L-Iditol hexaacetate

EMAIL THIS PAGE TO A FRIEND
302287 Aldrich

L-Iditol hexaacetate

99%

  • CAS Number 13443-46-4

  • Empirical Formula (Hill Notation) C18H26O12

  • Molecular Weight 434.39

  •  MDL number MFCD00064442

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?