• USA Home
  • 302902 - Hexamethylcyclotrisiloxane

EMAIL THIS PAGE TO A FRIEND
302902 Aldrich

Hexamethylcyclotrisiloxane

technical grade, 85%

Purchase

Properties

grade   technical grade
vapor density   1 (vs air)
InChI Key   HTDJPCNNEPUOOQ-UHFFFAOYSA-N
assay   85%
bp   134 °C(lit.)
mp   50-64 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 1325 4.1/PG 2
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?