• USA Home
  • 303364 - 4,7-Dihydro-2-phenyl-1,3-dioxepin

EMAIL THIS PAGE TO A FRIEND
303364 Sigma-Aldrich

4,7-Dihydro-2-phenyl-1,3-dioxepin

98%

  • CAS Number 2568-24-3

  • Empirical Formula (Hill Notation) C11H12O2

  • Molecular Weight 176.21

  •  MDL number MFCD00010646

  •  PubChem Substance ID 24858117

Purchase

Properties

assay   98%
refractive index   n20/D 1.543 (lit.)
bp   114 °C/3.5 mmHg(lit.)
density   1.11 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?