• USA Home
  • 303402 - Di-tert-butyl oxalate

EMAIL THIS PAGE TO A FRIEND
303402 Sigma-Aldrich

Di-tert-butyl oxalate

99%

  • CAS Number 691-64-5

  • Linear Formula (CH3)3COCOCOOC(CH3)3

  • Molecular Weight 202.25

  •  Beilstein/REAXYS Number 1774415

  •  EC Number 211-723-8

  •  MDL number MFCD00008806

  •  PubChem Substance ID 24858120

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C10 to C11, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   99%
mp   69-72 °C (lit.)
storage temp.   2-8°C
SMILES string   CC(C)(C)OC(=O)C(=O)OC(C)(C)C
InChI   1S/C10H18O4/c1-9(2,3)13-7(11)8(12)14-10(4,5)6/h1-6H3
InChI key   CIYGMWIAXRMHQS-UHFFFAOYSA-N

Description

General description

Di-tert-butyl oxalate is an ester.

Application

Di-tert-butyl oxalate was used in preparation of disodium salt of 2-[(dihydroxyphosphinyl) difluoromethyl] propenoic acid.

Packaging

5, 25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?