• USA Home
  • 303755 - Citral dimethyl acetal

EMAIL THIS PAGE TO A FRIEND
303755 Sigma-Aldrich

Citral dimethyl acetal

95%

Synonym: 3,7-Dimethyl-2,6-octadienal dimethyl acetal

Purchase

Properties

vapor density   6.8 (vs air)
vapor pressure   9 mmHg ( 102.7 °C)
description   mixture of cis and trans
InChI Key   ZSKAJFSSXURRGL-PKNBQFBNSA-N
assay   95%
refractive index   n20/D 1.454 (lit.)
bp   105-106 °C/10 mmHg(lit.)
density   0.89 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
2
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?