• USA Home
  • 303917 - 5-(4-Pyridyl)-2,7-nonadiene,mixture of isomers

EMAIL THIS PAGE TO A FRIEND
303917 Aldrich

5-(4-Pyridyl)-2,7-nonadiene,mixture of isomers

85%, technical grade

  • CAS Number 2057-34-3

  • Empirical Formula (Hill Notation) C14H19N

  • Molecular Weight 201.31

  •  MDL number MFCD00006438

Purchase

Properties

grade   technical grade
assay   85%
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?