• USA Home
  • 304441 - 4-Methyl-4-trichloromethyl-2,5-cyclohexadien-1-one

EMAIL THIS PAGE TO A FRIEND
304441

4-Methyl-4-trichloromethyl-2,5-cyclohexadien-1-one

99%

  • CAS Number 3274-12-2

  • Empirical Formula (Hill Notation) C8H7Cl3O

  • Molecular Weight 225.50

  •  MDL number MFCD00001590

Purchase

Properties

assay   99%
SMILES string   CC1(C=CC(=O)C=C1)C(Cl)(Cl)Cl
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?