• USA Home
  • 305170 - Methyl 7-oxooctadecanoate

EMAIL THIS PAGE TO A FRIEND
305170 Sigma-Aldrich

Methyl 7-oxooctadecanoate

97%

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   QTHPCXRUUSTYCU-UHFFFAOYSA-N
assay   97%
mp   50-52 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?