• USA Home
  • 306762 - α-D-Glucoheptonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
306762 Aldrich

α-D-Glucoheptonic acid sodium salt

97%

  • CAS Number 10094-62-9

  • Empirical Formula (Hill Notation) C7H17NaO9

  • Molecular Weight 268.19

  •  MDL number MFCD00013425

Purchase

Properties

assay   97%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?