• USA Home
  • 306827 - trans,trans-5-[4-(Dimethylamino)phenyl]-2,4-pentadienal

EMAIL THIS PAGE TO A FRIEND
306827 Sigma-Aldrich

trans,trans-5-[4-(Dimethylamino)phenyl]-2,4-pentadienal

99%

  • CAS Number 20432-36-4

  • Linear Formula (CH3)2NC6H4CH=CHCH=CHCHO

  • Molecular Weight 201.26

  •  MDL number MFCD00011579

Purchase

Properties

assay   99%
mp   155-156 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?