• USA Home
  • 307033 - 1,4-Cyclohexanedicarboxylic acid,mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
307033 Aldrich

1,4-Cyclohexanedicarboxylic acid,mixture of cis and trans

99%

Purchase

Properties

InChI Key   PXGZQGDTEZPERC-UHFFFAOYSA-N
assay   99%
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GU9060000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?