EMAIL THIS PAGE TO A FRIEND
307092

4-Methylphenetole

99%

Synonym: Ethyl p-tolyl ether

Purchase

Properties

assay   99%
SMILES string   CCOc1ccc(C)cc1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
158 °F
Flash Point(C) 
70 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?