• USA Home
  • 307769 - 2,3,5,6-Tetrafluoro-p-xylene

EMAIL THIS PAGE TO A FRIEND
307769 Sigma-Aldrich

2,3,5,6-Tetrafluoro-p-xylene

99%

Purchase

Properties

Related Categories Aryl, Aryl Fluorinated Building Blocks, Building Blocks, C7-C8, C8,
InChI Key   IWKPBYPUIPVYNZ-UHFFFAOYSA-N
assay   99%
bp   143-144 °C(lit.)
mp   35-37 °C (lit.)
storage temp.   2-8°C

Description

Application

2,3,5,6-Tetrafluoro-p-xylene was used in the synthesis of 1,4-di-α-bromomethyl-2,3,5,6-tetrafluorobenzene.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?