• USA Home
  • 307904 - 3-Ethoxy-4-methoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
307904

3-Ethoxy-4-methoxybenzyl alcohol

99%

  • CAS Number 147730-26-5

  • Empirical Formula (Hill Notation) C10H14O3

  • Molecular Weight 182.22

  •  MDL number MFCD00012291

Purchase

Properties

assay   99%
SMILES string   CCOc1cc(CO)ccc1OC
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?