EMAIL THIS PAGE TO A FRIEND
307998 Sigma-Aldrich

2-Ethylbenzenethiol

technical grade, 90%

Synonym: 2-Ethylthiophenol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
grade   technical grade
InChI Key   ABROBCBIIWHVNS-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.5682(lit.)
bp   203-205 °C(lit.)
density   1.025 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

General description

2-Ethylbenzenethiol is a strong mixed type inhibitor of ethylbenzene dehydrogenase.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
177.8 °F
Flash Point(C) 
81 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?