• USA Home
  • 308048 - 2-Furyl tetramethylphosphorodiamidate

EMAIL THIS PAGE TO A FRIEND
308048 Aldrich

2-Furyl tetramethylphosphorodiamidate

98%

  • CAS Number 105262-58-6

  • Empirical Formula (Hill Notation) C8H15N2O3P

  • Molecular Weight 218.19

  •  MDL number MFCD00012327

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?