• USA Home
  • 308447 - Sodium selenate decahydrate

EMAIL THIS PAGE TO A FRIEND
308447 Sigma-Aldrich

Sodium selenate decahydrate

technical grade

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Materials Science, Metal and Ceramic Science,
grade   technical grade
assay   85%
form   crystalline
mp   35 °C (dec.) (lit.)
density   1.6 g/mL at 25 °C
SMILES string   [Na+].[Na+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][Se]([O-])(=O)=O
InChI   1S/2Na.H2O4Se.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2
InChI key   TYVNFQRMDPFYPC-UHFFFAOYSA-L

Description

Packaging

50 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2630PSN1 6.1 / PGI
WGK Germany 
2
RTECS 
VS6720000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?