• USA Home
  • 309613 - (1R,exo,exo)-3-[N-(3,5-Dimethylphenyl)benzenesulfonamido]isoborneol

EMAIL THIS PAGE TO A FRIEND
309613 Sigma-Aldrich

(1R,exo,exo)-3-[N-(3,5-Dimethylphenyl)benzenesulfonamido]isoborneol

99%

Synonym: (1R,2S,3R)-(+)-3-[N-Benzenesulfonyl-N-(3,5-dimethylphenyl)amino]-2-bornanol

  • CAS Number 87420-26-6

  • Empirical Formula (Hill Notation) C24H31NO3S

  • Molecular Weight 413.57

  •  Beilstein/REAXYS Number 4715153

  •  MDL number MFCD00078990

Purchase

Properties

assay   99%
optical activity   [α]20/D +65°, c = 4 in chloroform
mp   122-125 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?