EMAIL THIS PAGE TO A FRIEND
310727 Sigma-Aldrich

Indophenol

Dye content 30 %

Synonym: Phenolindophenol

  • CAS Number 500-85-6

  • Empirical Formula (Hill Notation) C12H9NO2

  • Molecular Weight 199.21

  •  Beilstein/REAXYS Number 2095656

  •  EC Number 207-913-5

  •  MDL number MFCD00001621

Purchase

Properties

composition   Dye content, 30%
mp   >300 °C (lit.)
SMILES string   Oc1ccc(cc1)N=C2C=CC(=O)C=C2
InChI   1S/C12H9NO2/c14-11-5-1-9(2-6-11)13-10-3-7-12(15)8-4-10/h1-8,14H
InChI key   RSAZYXZUJROYKR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
GU5778000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I5763 Indophenol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?