EMAIL THIS PAGE TO A FRIEND
320633 Millipore

Cyclohexane

≥99.5%, PRA grade

  • CAS Number 110-82-7

  • Empirical Formula (Hill Notation) C6H12

  • Molecular Weight 84.16

  •  Beilstein/REAXYS Number 1900225

  •  EC Number 203-806-2

  •  MDL number MFCD00003814

  •  PubChem Substance ID 329753723

Purchase

Properties

Related Categories Amber Glass Bottles, PRA, Solvent Bottles, Solvent Packaging Options, Solvents More...
grade   PRA grade
vapor density   2.9 (vs air)
vapor pressure   168.8 mmHg ( 37.7 °C)
  77 mmHg ( 20 °C)
assay   ≥99.5%
autoignition temp.   500 °F
expl. lim.   9 %
evapn. residue   <0.0001%
halogenated residue (GC/ECD)   <10 ng/L (as heptachlor epoxide)
refractive index   n20/D 1.426 (lit.)
bp   80.7 °C (lit.)
mp   4-7 °C (lit.)
density   0.779 g/mL at 25 °C (lit.)
SMILES string   C1CCCCC1
InChI   1S/C6H12/c1-2-4-6-5-3-1/h1-6H2
InChI key   XDTMQSROBMDMFD-UHFFFAOYSA-N

Description

Application

Suitable for pesticide residue analysis

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
RIDADR 
UN1145 - class 3 - PG 2 - Cyclohexane
WGK Germany 
WGK 2
RTECS 
GU6300000
Flash Point(F) 
-0.4 °F - closed cup
Flash Point(C) 
-18.0 °C - closed cup
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?