• USA Home
  • 323721 - 2-(2-Propoxyethyl)pyridine

EMAIL THIS PAGE TO A FRIEND
323721 Aldrich

2-(2-Propoxyethyl)pyridine

99%

  • CAS Number 70644-45-0

  • Empirical Formula (Hill Notation) C10H15NO

  • Molecular Weight 165.23

  •  MDL number MFCD00006363

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?