EMAIL THIS PAGE TO A FRIEND
324027 Sigma-Aldrich

Ethyl acetate

PRA grade, ≥99.5%

Synonym: EtOAc

Purchase

Properties

Related Categories Amber Glass Bottles, Ethyl Acetate, PRA, Solvent by Type, Solvent Bottles,
grade   PRA grade
vapor density   3 (20 °C, vs air)
vapor pressure   73 mmHg ( 20 °C)
assay   ≥99.5%
autoignition temp.   801 °F
expl. lim.   2.2-11.5 %, 38 °F
impurities   ≤0.03% water
water
evapn. residue   <0.0001%
halogenated residue (GC/ECD)   <10 ng/L (as heptachlor epoxide)
refractive index   n20/D 1.3720 (lit.)
bp   76.5-77.5 °C (lit.)
  76.5-77.5 °C
mp   −84 °C (lit.)
solubility   water: soluble
density   0.902 g/mL at 25 °C (lit.)
SMILES string   CCOC(C)=O
InChI   1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
InChI key   XEKOWRVHYACXOJ-UHFFFAOYSA-N

Description

General description

Ethyl acetate is a good substitute solvent for diethyl ether, when it comes to formalin-ether sedimentation technique. It has also found functionality as an acyl receptor.

Application

It was used as an organic solvent, during the preparation of protein-loaded microparticles, which had a high microparticle yield.

Packaging

4×4 L in glass bottle

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1173 - class 3 - PG 2 - Ethyl acetate
WGK Germany 
1
RTECS 
AH5425000
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?