EMAIL THIS PAGE TO A FRIEND
324086 Aldrich

2-Propanol

99.9%, PRA grade

Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

  • CAS Number 67-63-0

  • Linear Formula (CH3)2CHOH

  • Molecular Weight 60.10

  •  Beilstein Registry Number 635639

  •  EC Number 200-661-7

  •  MDL number MFCD00011674

  •  PubChem Substance ID 329754189

Purchase

Properties

Related Categories Amber Glass Bottles, PRA, Solvent Bottles, Solvent Packaging Options, Solvents More...
grade   PRA grade
vapor density   2.1 (vs air)
vapor pressure   33 mmHg ( 20 °C)
  44 mmHg ( 25 °C)
InChI Key   KFZMGEQAYNKOFK-UHFFFAOYSA-N
assay   99.9%
autoignition temp.   750 °F
expl. lim.   2 %
impurities   ≤0.06% water
water
evapn. residue   <0.0002%
halogenated residue (GC/ECD)   <4 μg/L (as heptachlor epoxide)
refractive index   n20/D 1.377(lit.)
bp   82 °C(lit.)
mp   −89.5 °C(lit.)
density   0.785 g/mL at 25 °C(lit.)

Description

Application

Suitable for pesticide residue analysis

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1219 - class 3 - PG 2 - Isopropanol
WGK Germany 
1
RTECS 
NT8050000
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

The Pressure-Temperature Nomograph tool is an application of the Clausius-Clapeyron Equation, which assumes the heat of vaporization is a constant over a pressure range. Antoine´s Equation gets aroun...
Keywords: Evaporation, Vaporization

Properties of Solvents Table

a. Gant, R., LC/GC 10 (7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10 mmHg pressure 4. At l=589.32 nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?