• USA Home
  • 324434 - 3,3′-Methylenebis(α,α,α-trifluorotoluene)

EMAIL THIS PAGE TO A FRIEND
324434 Aldrich

3,3′-Methylenebis(α,α,α-trifluorotoluene)

97%

Synonym: 1,1′-Methylene-bis(3-[trifluoromethyl]benzene)

Purchase

Properties

assay   97%
mp   40-42 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?