• USA Home
  • 324841 - 2-Nitro-4-(trifluoromethyl)benzenesulfonamide

EMAIL THIS PAGE TO A FRIEND
324841

2-Nitro-4-(trifluoromethyl)benzenesulfonamide

98%

  • CAS Number 577-61-7

  • Empirical Formula (Hill Notation) C7H5F3N2O4S

  • Molecular Weight 270.19

  •  MDL number MFCD00013462

Purchase

Properties

assay   98%
SMILES string   NS(=O)(=O)c1ccc(cc1[N+]([O-])=O)C(F)(F)F
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?