EMAIL THIS PAGE TO A FRIEND
327840 Sigma-Aldrich

Phenylphosphine

98%

  • CAS Number 638-21-1

  • Linear Formula C6H5PH2

  • Molecular Weight 110.09

  •  Beilstein/REAXYS Number 741946

  •  EC Number 211-325-4

  •  MDL number MFCD00002094

Purchase

Properties

vapor density   3.79 (vs air)
vapor pressure   760 mmHg ( 160 °C)
InChI Key   RPGWZZNNEUHDAQ-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.578 (lit.)
bp   160 °C(lit.)
density   1.001 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2845 4.2
WGK Germany 
3
RTECS 
SZ2100000
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

674389 Phenylphosphine, ca. 10% weight in hexanes

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?