• USA Home
  • 327948 - 4-[(S)-(−)-2-Ethoxypropoxy]phenyl 4-(decyloxy)benzoate

EMAIL THIS PAGE TO A FRIEND
327948 Aldrich

4-[(S)-(−)-2-Ethoxypropoxy]phenyl 4-(decyloxy)benzoate

liquid crystal (Ferroelectric), 98%

  • CAS Number 103239-85-6

  • Empirical Formula (Hill Notation) C28H40O5

  • Molecular Weight 456.61

  •  MDL number MFCD00013231

Purchase

Properties

assay   98%
form   liquid crystal (Ferroelectric)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?