• USA Home
  • 328189 - 2-Butyl-1,3,2-dioxaborinane

EMAIL THIS PAGE TO A FRIEND
328189 Sigma-Aldrich

2-Butyl-1,3,2-dioxaborinane

98%

  • CAS Number 30169-71-2

  • Empirical Formula (Hill Notation) C7H15BO2

  • Molecular Weight 142.00

  •  MDL number MFCD00013363

Purchase

Properties

assay   98%
SMILES string   CCCCB1OCCCO1
InChI   1S/C7H15BO2/c1-2-3-5-8-9-6-4-7-10-8/h2-7H2,1H3
InChI key   UDMVQMMUSQGSDO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?