• USA Home
  • 328618 - 1-Bromo-2,3,4-trifluorobenzene

EMAIL THIS PAGE TO A FRIEND
328618 Sigma-Aldrich

1-Bromo-2,3,4-trifluorobenzene

99%

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   MUUAQFJJUGVBGB-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.487(lit.)
density   1.777 g/mL at 25 °C(lit.)

Description

Application

1-Bromo-2,3,4-trifluorobenzene was used in synthesis of 5-nitro-6-methylquinoline carboxylic acid.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?