• USA Home
  • 329509 - Nickel(II) acetylacetonate

EMAIL THIS PAGE TO A FRIEND
329509 Sigma-Aldrich

Nickel(II) acetylacetonate

technical grade, 90%

Synonym: 2,4-Pentanedione nickel(II) derivative, Ni(acac)2

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Nickel Catalysts More...
grade   technical grade
assay   90%
form   solid
mp   230 °C (dec.) (lit.)
SMILES string   CC(=O)\C=C(\C)O[Ni]O\C(C)=C/C(C)=O
InChI   1S/2C5H8O2.Ni/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;
InChI key   BMGNSKKZFQMGDH-FDGPNNRMSA-L
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SA2100000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

283657 Nickel(II) acetylacetonate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?