• USA Home
  • 331236 - trans-1,4-Cyclohexanedicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
331236 Sigma-Aldrich

trans-1,4-Cyclohexanedicarboxylic acid

93%

Purchase

Properties

assay   93%
mp   >300 °C (lit.)
SMILES string   OC(=O)[C@@H]1CC[C@H](CC1)C(O)=O
InChI   1S/C8H12O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-
InChI key   PXGZQGDTEZPERC-IZLXSQMJSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
WGK 3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?