• USA Home
  • 331414 - trans-Dichlorobis(triethylphosphine)platinum(II)

EMAIL THIS PAGE TO A FRIEND
331414 Sigma-Aldrich

trans-Dichlorobis(triethylphosphine)platinum(II)

98%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Platinum Catalysts More...
Quality Level   100
assay   98%
reaction suitability   core: platinum
  reagent type: catalyst
mp   141-143 °C (lit.)
SMILES string   Cl[Pt]Cl.CCP(CC)CC.CCP(CC)CC
InChI   1S/2C6H15P.2ClH.Pt/c2*1-4-7(5-2)6-3;;;/h2*4-6H2,1-3H3;2*1H;/q;;;;+2/p-2
InChI key   WPWLTKRUFHHDLP-UHFFFAOYSA-L

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?