• USA Home
  • 331619 - (S)-(−)-1-Phenyl-1-decanol

EMAIL THIS PAGE TO A FRIEND
331619 Sigma-Aldrich

(S)-(−)-1-Phenyl-1-decanol

95%

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   MRZGGLIGGZHSBZ-INIZCTEOSA-N
assay   95%
optical activity   [α]20/D −14.4°, neat
refractive index   n20/D 1.497(lit.)
bp   128 °C/0.35 mmHg(lit.)
density   0.922 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?