• USA Home
  • 331627 - N-Phenylthio-ε-caprolactam

EMAIL THIS PAGE TO A FRIEND
331627 Aldrich

N-Phenylthio-ε-caprolactam

98%

  • CAS Number 36452-23-0

  • Empirical Formula (Hill Notation) C12H15NOS

  • Molecular Weight 221.32

  •  MDL number MFCD00074944

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?