• USA Home
  • 332194 - Cyclohexylsuccinic acid

EMAIL THIS PAGE TO A FRIEND
332194 Sigma-Aldrich

Cyclohexylsuccinic acid

96%

Purchase

Properties

assay   96%
mp   145-147 °C (lit.)
SMILES string   OC(=O)CC(C1CCCCC1)C(O)=O
InChI   1S/C10H16O4/c11-9(12)6-8(10(13)14)7-4-2-1-3-5-7/h7-8H,1-6H2,(H,11,12)(H,13,14)
InChI key   FROUMWCGMNOSBK-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?