• USA Home
  • 332267 - 3-O-Acetyl-6-O-benzoyl-5-O-(methylsulfonyl)-1,2-O-isopropylidene-α-D-glucofuranose

EMAIL THIS PAGE TO A FRIEND
332267 Sigma-Aldrich

3-O-Acetyl-6-O-benzoyl-5-O-(methylsulfonyl)-1,2-O-isopropylidene-α-D-glucofuranose

97%

  • CAS Number 102029-58-3

  • Empirical Formula (Hill Notation) C19H24O10S

  • Molecular Weight 444.45

  •  MDL number MFCD00074967

Purchase

Properties

assay   97%
optical activity   [α]25/D −9°, c = 1 in chloroform
mp   128-130 °C (lit.)
SMILES string   CC(=O)O[C@H]1[C@H](O[C@@H]2OC(C)(C)O[C@H]12)[C@@H](COC(=O)c3ccccc3)OS(C)(=O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?