• USA Home
  • 332496 - Cinnoline hydrochloride

EMAIL THIS PAGE TO A FRIEND
332496 Aldrich

Cinnoline hydrochloride

98%

  • CAS Number 5949-24-6

  • Empirical Formula (Hill Notation) C8H6N2 · HCl

  • Molecular Weight 166.61

  •  MDL number MFCD00074974

  •  PubChem Substance ID 24859901

Purchase

Properties

Related Categories C, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   JQBSTRWZKVHRHE-UHFFFAOYSA-N
assay   98%
mp   156-158 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Milli-Q® Water Purification Solutions

Biomedical Applications
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?