• USA Home
  • 333166 - 1,4,8,11-Tetraazacyclotetradecane-5,7-dione

EMAIL THIS PAGE TO A FRIEND
333166 Sigma-Aldrich

1,4,8,11-Tetraazacyclotetradecane-5,7-dione

99%

Synonym: Dioxocyclam

  • CAS Number 63972-19-0

  • Empirical Formula (Hill Notation) C10H20N4O2

  • Molecular Weight 228.29

  •  Beilstein/REAXYS Number 1214926

  •  MDL number MFCD00075005

  •  PubChem Substance ID 24860107

  •  NACRES NA.23

Purchase

Properties

Related Categories Amine Monomers, Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Materials Science,
assay   99%
mp   172-176 °C (lit.)
SMILES string   O=C1CC(=O)NCCNCCCNCCN1
InChI   1S/C10H20N4O2/c15-9-8-10(16)14-7-5-12-3-1-2-11-4-6-13-9/h11-12H,1-8H2,(H,13,15)(H,14,16)
InChI key   BGVLBVASHIQNIO-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3259PSN2 8 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?