• USA Home
  • 334014 - 3-(Phenylsulfonyl)propionitrile

EMAIL THIS PAGE TO A FRIEND
334014 Sigma-Aldrich

3-(Phenylsulfonyl)propionitrile

98%

  • CAS Number 10154-75-3

  • Empirical Formula (Hill Notation) C9H9NO2S

  • Molecular Weight 195.24

  •  MDL number MFCD00047798

Purchase

Properties

assay   98%
SMILES string   O=S(=O)(CCC#N)c1ccccc1
InChI   1S/C9H9NO2S/c10-7-4-8-13(11,12)9-5-2-1-3-6-9/h1-3,5-6H,4,8H2
InChI key   OVZPZPVSUAKTCM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?