• USA Home
  • 334154 - Allyl 1-benzotriazolyl carbonate

EMAIL THIS PAGE TO A FRIEND
334154 Sigma-Aldrich

Allyl 1-benzotriazolyl carbonate

95%

Synonym: 1-(Allyloxycarbonyloxy)-1H-benzotriazole

  • CAS Number 102423-16-5

  • Empirical Formula (Hill Notation) C10H9N3O3

  • Molecular Weight 219.20

  •  MDL number MFCD00075024

Purchase

Properties

assay   95%
mp   110-113 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?