• USA Home
  • 335096 - Nitrosyltris(triphenylphosphine)rhodium(I)

EMAIL THIS PAGE TO A FRIEND
335096 Sigma-Aldrich

Nitrosyltris(triphenylphosphine)rhodium(I)

97%

Purchase

Properties

assay   97%
mp   147 °C (dec.) (lit.)
SMILES string   [Rh]N=O.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?