• USA Home
  • 335371 - Bismuth(III) oxyiodide

EMAIL THIS PAGE TO A FRIEND
335371 Aldrich

Bismuth(III) oxyiodide

Synonym: Bismuth(III) iodide oxide

Purchase

Properties

Related Categories Bismuth Catalysts, Catalysis and Inorganic Chemistry, Chemical Synthesis More...
InChI Key   YPMZHGJQYGMKBQ-UHFFFAOYSA-M
density   7.92 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?