• USA Home
  • 335797 - 3,5-Dibromoanthranilic acid

EMAIL THIS PAGE TO A FRIEND
335797 Sigma-Aldrich

3,5-Dibromoanthranilic acid

98%

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   98%
application(s)   peptide synthesis: suitable
mp   235-236 °C (lit.)
SMILES string   Nc1c(Br)cc(Br)cc1C(O)=O
InChI   1S/C7H5Br2NO2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,10H2,(H,11,12)
InChI key   WNABMWFLKQEGCP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?